Specifications
3'Primer ModificationNone
5'Primer ModificationNone
Concentration20X
FormatTube
Gene SymbolRPPH1
Green FeaturesSustainable packaging
Internal Probe ModificationTAMRA™ Quencher (3'), VIC™ (5')
No. of Reactions3000 Reactions
Product LineTaqMan™
Product TypeConfigurable Assay
Purification MethodHPLC
Purity or Quality GradeRNase-Free, DNase-Free
Quantity3,000 reactions
Shipping ConditionAmbient Temperature
SpeciesHuman
Sufficient For3000 Reactions
TargetRNase P
Detection MethodPrimer-probe
FormFrozen
GC-Rich PCR PerformanceLow
Label or DyeVIC
PCR MethodqPCR
Reaction SpeedStandard
Unit Size3,000 reactions