Thermo Scientific Chemicals

Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate, 97%, Thermo Scientific Chemicals

Catalog number: 440360050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate, 97%, Thermo Scientific Chemicals

Catalog number: 440360050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 440360050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
164298-25-3
IUPAC Name
1-[fluoro(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
Molecular Formula
C9H16F7N2P
InChI Key
MNJUGQKOHJQOCK-UHFFFAOYSA-N
SMILES
F[P-](F)(F)(F)(F)F.FC(N1CCCC1)=[N+]1CCCC1
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=96.0 %
Elemental Analysis
32.82 to 35.55 % (C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text