Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate, 97%, Thermo Scientific Chemicals
Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fluoro-N,N,N',N'-bis(tetramethylene)formamidinium hexafluorophosphate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 440360250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS164298-25-3
IUPAC Name1-[fluoro(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC9H16F7N2P
InChI KeyMNJUGQKOHJQOCK-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.FC(N1CCCC1)=[N+]1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Elemental Analysis32.82 to 35.55 % (C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only