Di-tert-butyl hydrazodiformate, 97%, Thermo Scientific Chemicals
Di-tert-butyl hydrazodiformate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-tert-butyl hydrazodiformate, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 440420100
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
Add to cart
Di-tert-butyl hydrazodiformate, 97%, Thermo Scientific Chemicals
Catalog number440420100
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS16466-61-8
IUPAC NameN'-[(tert-butoxy)carbonyl](tert-butoxy)carbohydrazide
Molecular FormulaC10H20N2O4
InChI KeyTYSZETYVESRFNT-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NNC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only