Applied Biosystems™

TaqMan™ OpenArray™ Genotyping Master Mix

Catalog number: 4404846
Have Questions?
Applied Biosystems™

TaqMan™ OpenArray™ Genotyping Master Mix

Catalog number: 4404846
Have Questions?
Catalog Number
4404846
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
44048461 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This 2X concentrated mix is optimized for SNP genotyping using TaqMan™ OpenArray™ SNP Genotyping Plates.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
PCR Method
qPCR
Polymerase
AmpliTaq Gold DNA Polymerase
Reaction Speed
Standard
Technique
SNP Genotyping (Real-Time PCR-Based)
For Use With (Equipment)
OpenArray™
Genotyping Target
Drug Metabolizing Enzymes, SNPs (Known)
Passive Reference Dye
ROX (Pre-mixed)
Product Line
OpenArray™, TaqMan™
Product Type
Genotyping Master Mix
Shipping Condition
Wet Ice
For Use With (Application)
Genotyping
Concentration
2X
Fidelity (vs. Taq)
2 X
Hot Start
Built-In Hot Start
Sample Type
DNA

Contents & Storage

Includes TaqMan™ OpenArray™ Genotyping Master Mix, with AmpliTaq Gold™ DNA Polymerase UP (Ultra Pure), dNTPs, and optimized mix components. This pack contains one 5 mL bottle, sufficient for 32 plates. Store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text