Thermo Scientific Chemicals

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%, Thermo Scientific Chemicals

Catalog number: 440521000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride, 98%, Thermo Scientific Chemicals

Catalog number: 440521000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 440521000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4568-71-2
IUPAC Name
3-benzyl-5-(2-hydroxyethyl)-4-methyl-1,3-thiazol-3-ium chloride
Molecular Formula
C13H16ClNOS
InChI Key
IWSVLBKHBJGMAA-UHFFFAOYSA-M
SMILES
[Cl-].CC1=C(CCO)SC=[N+]1CC1=CC=CC=C1
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
141°C to 147°C
Elemental Analysis
56.52 to 59.32 % C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text