Chemical Identifiers
CAS132327-80-1
IUPAC Name2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-[(triphenylmethyl)carbamoyl]butanoic acid
Molecular FormulaC39H34N2O5
InChI KeyWDGICUODAOGOMO-UHFFFAOYNA-N
SMILESOC(=O)C(CCC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View more