Thermo Scientific Chemicals

Xylene cyanol FF, ∽75%, Thermo Scientific Chemicals

Catalog number: 440700500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Xylene cyanol FF, ∽75%, Thermo Scientific Chemicals

Catalog number: 440700500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 440700500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2650-17-1
IUPAC Name
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate
Molecular Formula
C25H27N2NaO6S2
InChI Key
VVLFAAMTGMGYBS-KRQUPCAFSA-M
SMILES
[Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O
Loss on drying
=<5 % (105°C)
Appearance (Color)
Blue-grey to green
Appearance (Form)
Powder
UV Lambda max
612 to 616 nm
Dye content
>=70 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text