Thermo Scientific Chemicals

4,5-Dimethoxy-2-nitrobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 440770010
1 g, Each
Thermo Scientific Chemicals

4,5-Dimethoxy-2-nitrobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: 440770010
1 g, Each
Quantity
Catalog number: 440770010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
53413-67-5
IUPAC Name
1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene
Molecular Formula
C9H10BrNO4
InChI Key
UEKFEYNZISYRRH-UHFFFAOYSA-N
SMILES
COC1=CC(CBr)=C(C=C1OC)[N+]([O-])=O
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or needles
HPLC
>=96.0 %
Appearance (Color)
Yellow to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text