MicroSEQ™ ID Sequencing Clean-up Plates
Product Image
Applied Biosystems™
MicroSEQ™ ID Sequencing Clean-up Plates
The MicroSEQ™ ID Sequencing Clean-up Plates are used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel filtration matrixRead more
Have Questions?
Catalog NumberQuantity
44082272 x 96-well plates
Catalog number 4408227
Price (USD)
330.00
Each
Add to cart
Quantity:
2 x 96-well plates
Price (USD)
330.00
Each
Add to cart
The MicroSEQ™ ID Sequencing Clean-up Plates are used for post-sequencing clean-up in the MicroSEQ™ ID workflow. The gel filtration matrix used in the microplate cartridges provides a convenient and simple way to remove dye terminators, dNTPs, and salts, as well as residual primers from cycle sequencing reactions.
The MicroSEQ™ ID Sequencing Clean-up Plates can be used downstream of the following sequencing kits:
  • MicroSEQ™ 500 16S rDNA Sequencing Kit
  • MicroSEQ™ Full Gene 16S rDNA Sequencing Kit
  • MicroSEQ™ D2 rDNA Fungal Sequencing Kit
This product provides two 96-well microplate cartridges ideal for high throughput microbial identification workflows. The end products can be directly used for sequencing on any Applied Biosystems™ genetic analyzers.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)3500xL Genetic Analyzer, MicroSEQ™ ID, 3500 Genetic Analyzer
Product TypeMicroplate
Quantity2 x 96-well plates
Shipping ConditionWet Ice
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).
Share catalog number, name or link