Chemical Identifiers
CAS3674-06-4
IUPAC Name2,5-dioxopyrrolidin-1-yl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
Molecular FormulaC18H22N2O6
InChI KeyNHUCANAMPJGMQL-ZDUSSCGKSA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
View more