N-TIPS pyrrole-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
N-TIPS pyrrole-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-TIPS pyrrole-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 441020050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS365564-11-0
IUPAC Name3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[tris(propan-2-yl)silyl]-1H-pyrrole
Molecular FormulaC19H36BNO2Si
InChI KeyGWFIZBYDIHGZRJ-UHFFFAOYSA-N
SMILESCC(C)[Si](C(C)C)(C(C)C)N1C=CC(=C1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or solid
GC>=94.0 %
Infrared spectrumConforms
Appearance (Color)White to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only