Thermo Scientific Chemicals

(R)-N-BOC-α-Ethylalanine, 98%, 98% ee, Thermo Scientific Chemicals

Catalog number: 441065000
500 mg, Each
Thermo Scientific Chemicals

(R)-N-BOC-α-Ethylalanine, 98%, 98% ee, Thermo Scientific Chemicals

Catalog number: 441065000
500 mg, Each
Quantity
Catalog number: 441065000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
123254-58-0
IUPAC Name
2-{[(tert-butoxy)carbonyl]amino}-2-methylbutanoic acid
Molecular Formula
C10H19NO4
InChI Key
SHZXLTCEPXVCSV-SNVBAGLBSA-N
SMILES
CCC(C)(NC(=O)OC(C)(C)C)C(O)=O
Appearance (Color)
White to light yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Enantiomeric excess
>=98 % (HPLC)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text