Thermo Scientific Chemicals

(R)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific Chemicals

Catalog number: 441145000
500 mg, Each
Thermo Scientific Chemicals

(R)-N-FMOC-α-Methylvaline, 98%, 98% ee, Thermo Scientific Chemicals

Catalog number: 441145000
500 mg, Each
Quantity
Catalog number: 441145000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
616867-28-8
IUPAC Name
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-2,3-dimethylbutanoic acid
Molecular Formula
C21H23NO4
InChI Key
AWEZXIRZNQCCNN-OAQYLSRUSA-N
SMILES
CC(C)C(C)(NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
HPLC
>=97.5 %
Residual solvents
=<10 % MTBE
Specific optical rotation
-7.5° to -9.5° (20°C, 589 nm) (c=1 in CHCl3)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text