Thermo Scientific Chemicals

(S)-alpha-Methyltryptophan hemihydrate, 98%, 98% ee, Thermo Scientific Chemicals

Catalog number: 441201000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

(S)-alpha-Methyltryptophan hemihydrate, 98%, 98% ee, Thermo Scientific Chemicals

Catalog number: 441201000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 441201000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16709-25-4
IUPAC Name
bis(2-amino-3-(1H-indol-3-yl)-2-methylpropanoic acid) hydrate
Molecular Formula
C24H30N4O5
InChI Key
BZCFHAYHUKOGKE-NMFAMCKASA-N
SMILES
O.CC(N)(CC1=CNC2=CC=CC=C12)C(O)=O.CC(N)(CC1=CNC2=CC=CC=C12)C(O)=O
Appearance (Form)
Powder
Specific optical rotation
-14° to -18° (20°C, 589 nm) (c=0.9 in water)
Infrared spectrum
Conforms
Enantiomeric excess
>=98.0 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text