You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
Shop All Products
Clinical Analyzers and Instruments
DNA Sequencing Systems
Septa for 384-Well Plates, for 3500/SeqStudio™ Flex
Applied Biosystems™
Septa for 384-Well Plates, for 3500/SeqStudio™ Flex
Catalog number:
4412520
Related applications:
Sanger Sequencing
Technical Support
Customer Service
Applied Biosystems™
Septa for 384-Well Plates, for 3500/SeqStudio™ Flex
Catalog number:
4412520
Related applications:
Sanger Sequencing
Technical Support
Customer Service
Catalog Number
4412520
Unit Size
20 each
Price (TWD)
***
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (TWD)
Quantity
4412520
20 each
***
Save to list
Add to cart
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
These septa for 384-well plates are for use with Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format
384-well Plate
For Use With (Equipment)
3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
Product Type
Septa Plate
Shipping Condition
Room Temperature
Contents & Storage
This product contains 20 septa.
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search