Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)palladium(0), 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 441400050
5 g, Each
Thermo Scientific Chemicals

Tetrakis(triphenylphosphine)palladium(0), 99.9%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 441400050
5 g, Each
Quantity
Catalog number: 441400050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14221-01-3
IUPAC Name
tetrakis(triphenylphosphane) palladium
Molecular Formula
C72H60P4Pd
InChI Key
NFHFRUOZVGFOOS-UHFFFAOYSA-N
SMILES
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Color)
Yellow to yellow-green to orange
Palladium (Pd)
8.8 to 9.6 %
Total trace metal impurities
=<0.1 %
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text