N-Methylcarbazole-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
N-Methylcarbazole-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Methylcarbazole-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 441440010
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
97.40 
Save 14.75 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
97.40 
Save 14.75 (15%)
Add to cart
N-Methylcarbazole-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog number441440010
Price (USD)/ Each
82.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
97.40 
Save 14.75 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name9-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Molecular FormulaC19H22BNO2
InChI KeyAVCKYKVGQYBADD-UHFFFAOYSA-N
SMILESCN1C2=C(C=CC=C2)C2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol)307.20
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Melting point147°C to 154°C
HPLC>=96.0 %
Appearance (Color)Light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only