2-Amino-4-chloropyridine, 97%, Thermo Scientific Chemicals
2-Amino-4-chloropyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-4-chloropyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 441460250
Price (USD)/ Each
214.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
214.00
Add to cart
2-Amino-4-chloropyridine, 97%, Thermo Scientific Chemicals
Catalog number441460250
Price (USD)/ Each
214.00
-
Add to cart
Chemical Identifiers
CAS19798-80-2
IUPAC Name2-amino-4-chloropyridin-1-ium
Molecular FormulaC5H6ClN2
InChI KeyRQMWVVBHJMUJNZ-UHFFFAOYSA-O
SMILESNC1=CC(Cl)=CC=[NH+]1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)White to light brown
Infrared spectrumConforms
Melting point127°C to 134°C
Appearance (Form)Solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only