2-Phenylethylamine hydrochloride, 98%, Thermo Scientific Chemicals
2-Phenylethylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenylethylamine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number 441910250
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
IUPAC Namehydrogen 2-phenylethan-1-amine chloride
Molecular FormulaC8H12ClN
InChI KeySKHIBNDAFWIOPB-UHFFFAOYSA-N
SMILES[H+].[Cl-].NCCC1=CC=CC=C1
Molecular Weight (g/mol)157.64
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration Argentometric>=97.5 to =<102.5 %
Infrared spectrumConforms
Appearance (Form)Powder or solid
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only