Thermo Scientific Chemicals

L-Aspartic acid β-benzyl ester, 98%, Thermo Scientific Chemicals

Catalog number: 441930050
5 g, Each
Thermo Scientific Chemicals

L-Aspartic acid β-benzyl ester, 98%, Thermo Scientific Chemicals

Catalog number: 441930050
5 g, Each
Quantity
Catalog number: 441930050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2177-63-1
IUPAC Name
2-amino-4-(benzyloxy)-4-oxobutanoic acid
Molecular Formula
C11H13NO4
InChI Key
VGALFAWDSNRXJK-UHFFFAOYNA-N
SMILES
NC(CC(=O)OCC1=CC=CC=C1)C(O)=O
Appearance (Form)
Powder
Specific optical rotation
+26° to +30° (20°C, 589 nm) (c=1 in dil HCl)
Appearance (Color)
White
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text