BOC-L-Tyrosine methyl ester, 97%, Thermo Scientific Chemicals
BOC-L-Tyrosine methyl ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

BOC-L-Tyrosine methyl ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 442020050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Namemethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Molecular FormulaC15H21NO5
InChI KeyNQIFXJSLCUJHBB-STGVRZAANA-N
SMILESCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C
Molecular Weight (g/mol)295.34
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Form)Powder
Appearance (Color)White to light yellow
Infrared spectrumConforms
Specific optical rotation+48° to +52° (20°C, 589 nm) (c=1 in CHCl3)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only