Sodium methanesulfonate, 99%
Sodium methanesulfonate, 99%
Sodium methanesulfonate, 99%
Sodium methanesulfonate, 99%
Thermo Scientific Chemicals

Sodium methanesulfonate, 99%

CAS: 2386-57-4 | CH4NaO3S | 118.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Catalog number 442115000
Price (USD)
372.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
372.00
Each
Add to cart
Chemical Identifiers
CAS2386-57-4
IUPAC Namemethanesulfonic acid sodium
Molecular FormulaCH4NaO3S
InChI KeyDWNWZFAKFWMPTI-UHFFFAOYSA-N
SMILES[Na].CS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Water=<1 %
Free acid=<1 %
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link