Thermo Scientific Chemicals

Pyridine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: 442132500
250 g, Each
Thermo Scientific Chemicals

Pyridine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog number: 442132500
250 g, Each
Quantity
Catalog number: 442132500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18820-82-1
IUPAC Name
hydrogen pyridine bromide
Molecular Formula
C5H6BrN
InChI Key
BBFCIBZLAVOLCF-UHFFFAOYSA-N
SMILES
[H+].[Br-].C1=CC=NC=C1
Appearance (Form)
Powder or solid
Water
=<0.5 %
Infrared spectrum
Conforms
Titration Argentometric
>=97.5 %
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text