Thermo Scientific Chemicals

(S)-(+)-2-Phenylglycine methyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 442190100
10 g, Each
Thermo Scientific Chemicals

(S)-(+)-2-Phenylglycine methyl ester hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 442190100
10 g, Each
Quantity
Catalog number: 442190100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15028-39-4
IUPAC Name
methyl 2-amino-2-phenylacetate hydrochloride
Molecular Formula
C9H12ClNO2
InChI Key
DTHMTBUWTGVEFG-UHFFFAOYNA-N
SMILES
Cl.COC(=O)C(N)C1=CC=CC=C1
Appearance (Form)
Crystalline powder
Specific optical rotation
+131° to +137° (20°C, 589 nm) (c=1,MeOH)
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Loss on drying
=<1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text