Thermo Scientific Chemicals

4-Benzyloxybromobenzene, 97%, Thermo Scientific Chemicals

Catalog number: 442460100
10 g, Each
Thermo Scientific Chemicals

4-Benzyloxybromobenzene, 97%, Thermo Scientific Chemicals

Catalog number: 442460100
10 g, Each
Quantity
Catalog number: 442460100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6793-92-6
IUPAC Name
1-(benzyloxy)-4-bromobenzene
Molecular Formula
C13H11BrO
InChI Key
OUQSGILAXUXMGI-UHFFFAOYSA-N
SMILES
BrC1=CC=C(OCC2=CC=CC=C2)C=C1
Infrared spectrum
Conforms
Melting point
59°C to 64°C
GC
>=96.0 %
Appearance (Color)
White to light brown
Appearance (Form)
Powder or needles

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text