Applied Biosystems™

DS-36 Matrix Standard (Dye Set J6)

Catalog number: 4425042
Have Questions?
Applied Biosystems™

DS-36 Matrix Standard (Dye Set J6)

Catalog number: 4425042
Have Questions?
Catalog Number
4425042
Unit Size
8 runs
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
44250428 runs
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The DS-36 Matrix Standard (Dye Set J6) is used to perform the spectral calibration required to analyze 6-FAM, VIC, NED, SID, TAZ, and LIZ dye-labeled DNA fragments on the Applied Biosystems SeqStudio, 3130, 3500, and 3730 series systems. The data collection software for these instruments uses the multi-component matrix to automatically analyze the 6 differently colored fluorescent dye-labeled samples in a single capillary. The DS-36 Matrix Standard (Dye Set J6) contains 6 specific sizes of a unique fluorescent dye label.

Specifications

Software Type
Data Collection Software
Starting Material
DNA
For Use With (Equipment)
3500xL Genetic Analyzer, 3130xl Genetic Analyzer, 3130 Genetic Analyzer, 3500 Genetic Analyzer
Label or Dye
6-FAM, LIZ, NED, SID, TAZ, VIC
Product Type
DS-36 Matrix Standard
Quantity
8 Runs
Shipping Condition
Wet Ice
Unit Size
8 runs

Contents & Storage

Store in refrigerator (2–8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text