Thermo Scientific Chemicals

Acetoacetamide, 97%, Thermo Scientific Chemicals

Catalog number: 442541000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Acetoacetamide, 97%, Thermo Scientific Chemicals

Catalog number: 442541000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 442541000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5977-14-0
IUPAC Name
3-oxobutanamide
Molecular Formula
C4H7NO2
InChI Key
GCPWJFKTWGFEHH-UHFFFAOYSA-N
SMILES
CC(=O)CC(N)=O
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
52°C to 58°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text