Applied Biosystem® ready-to-use MeltDoctor™ HRM Calibration Plates contain all the components required for pure dye and HRM calibration, reducing theRead more
Have Questions?
Catalog number 4425618
Price (USD)/ 1 x 96 well plate
751.65
Online exclusive
808.00
Save 56.35 (7%)
-
Add to cart
Price (USD)/ 1 x 96 well plate
751.65
Online exclusive
808.00
Save 56.35 (7%)
Add to cart
MeltDoctor™ HRM Calibration Plate, Fast 96-well
Catalog number4425618
Price (USD)/ 1 x 96 well plate
751.65
Online exclusive
808.00
Save 56.35 (7%)
-
Add to cart
Applied Biosystem® ready-to-use MeltDoctor™ HRM Calibration Plates contain all the components required for pure dye and HRM calibration, reducing the complexity of getting started with HRM experimentation. These plates provide thermal and optical calibration required for accurate and reproducible discrimination between melt curves.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format96-well Plate
Calibrated DyeHRM (High Resolution Melting) Dye
For Use With (Equipment)7500 Fast System, 7900HT Fast System