1x1 image pixel for data collection
Genetic Analyzer Upgrade Kit (3500 to 3500xL)
Genetic Analyzer Upgrade Kit (3500 to 3500xL)
Applied Biosystems™
Genetic Analyzer Upgrade Kit (3500 to 3500xL)
This kit allows you to upgrade your 8 capillary 3500 Genetic Analyzer to a 24 capillary 3500xL expanding the capabilitiesRead more
Have Questions?
Catalog NumberQuantity
44264791 Kit
Catalog number 4426479
Price (USD)
-
Request A Quote
Quantity:
1 Kit
This kit allows you to upgrade your 8 capillary 3500 Genetic Analyzer to a 24 capillary 3500xL expanding the capabilities of your instrument. You’ll be able to take advantage of a dramatic increase in throughput and in overall laboratory productivity. The upgrade is simple and completed within a day. It does not involve any changes to the computer or software so you will not need to re-verify these components of your system.
• 8 capillary system can easily be upgraded to a 24-capillary system
• New single-line 505 nm, solid-state long-life laser—utilizes a standard power supply; requires no heat-removal ducting
• Powerful, integrated data collection and primary analysis software provides real-time assessment of data quality
• Radio Frequency Identification (RFID) technology tracks key consumables data and records administrative information
• Advanced multiplexing capabilities for DNA fragment analysis with up to six unique dyes
• Unrivaled application flexibility—one array and one polymer are used for most applications
• Simple setup, operation and maintenance—the easiest-to-run, easiest-to-own DNA sequencer to date
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)3500 Genetic Analyzer
Product TypeGenetic Analyzer Upgrade Kit
Quantity1 Kit
System Upgrade3500 to 3500XL
FormatKit
Unit SizeEach
Contents & Storage
This upgrade comes with a starter kit.
Frequently asked questions (FAQs)
Is there a way to automatically upload my data into the Thermo Fisher Cloud app directly from the Applied Biosystems 3500 Genetic Analyzer, instead of using Data Collection Software?
If you are using the most recent version of Data Collection Software (v3.3), there is a way to automatically upload your data into the Thermo Fisher Cloud app from the instrument. When a run is complete, the data is stored in the results group folder that is specified in the plate record. The data will be automatically uploaded to your account if your User ID is the active ID. Results are saved in your account folders named by the plate and injections, not in the results group folder. Please go to page 43 of the 3500/3500xL Genetic Analyzer User Guide for Data Collection Software v3.3 (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/100079380_3500v3_3RUO_UG.pdf) for details regarding the Thermo Fisher Cloud app.
If you are using Data Collection Software v3.1, you would have to manually upload your data into the Thermo Fisher Cloud app from your data folder on the computer.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Instruments Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link