TaqMan™ Pri-miRNA Assay
TaqMan™ Pri-miRNA Assay
Applied Biosystems™

TaqMan™ Pri-miRNA Assay

Green features
TaqMan Pri-miRNA Assays quantitate primary microRNA (pri-miRNA) transcripts, the most upstream RNA molecules in the microRNA biogenesis pathway. They areRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4427012S (360 reactions), made to order
4427013M (750 reactions), made to order
4427014L (2900 reactions), made to order
3 Options
Catalog number 4427012
Price (USD)/ S (360 reactions), made to order
415.65
Online exclusive
442.00 
Save 26.35 (6%)
Order now
Quantity:
S (360 reactions), made to order

TaqMan Pri-miRNA Assays quantitate primary microRNA (pri-miRNA) transcripts, the most upstream RNA molecules in the microRNA biogenesis pathway. They are designed using our state-of-the-art bioinformatics pipeline, delivering gold-standard assay performance and data quality for pri-miRNA studies.

Highly specific—quantitiate miRNA transcription from a single genomic locus
Fast, simple, and scalable—two-step RT-qPCR provides high-quality results
A new perspective—measure miRNA expression at the gene level

TaqMan Pri-miRNA Assays apply the same high sensitivity, superior specificity, and wide linear dynamic range for which all TaqMan genomic assays are renowned to the detection of pri-miRNA transcripts. As a result of their high sensitivity, TaqMan Pri-miRNA Assays require as little as 1 ng of sample input to quantify expression from moderate- to high-expression miRNA loci.

Design and selection
Since all pri-miRNA transcripts have not been mapped, these assays are designed within close proximity to each stem-loop sequence represented in the Sanger miRBase sequence repository. This ensures accurate measurement of the transcript of interest. Each publicly available stem-loop can be matched to a TaqMan Pri-miRNA Assay and TaqMan MicroRNA Assay, enabling the RNA sequences produced at either end of the miRNA maturation pathway to be quantified independently.

Pre-formulated assay (20X; 60X mix)
• 2 unlabeled PCR primers (900 nM each final 1X reaction concentration)
• 1 FAM dye-labeled TaqMan MGB probe (250 nM final 1X reaction concentration)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast
Green FeaturesSustainable packaging
Product LineTaqMan™
Product TypePri-miRNA Assay
QuantityS (360 reactions), made to order
Shipping ConditionRoom Temperature
Sufficient For360 Reactions
Concentration20X
Fidelity (vs. Taq)60 X
No. of Reactions360 reactions
Unit SizeS (360 reactions), made to order
Contents & Storage
1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.