Thermo Scientific Chemicals

N-BOC-1,3-diaminopropane, 97%, Thermo Scientific Chemicals

Catalog number: 442730050
5 mL, Glass Bottle, Each
Thermo Scientific Chemicals

N-BOC-1,3-diaminopropane, 97%, Thermo Scientific Chemicals

Catalog number: 442730050
5 mL, Glass Bottle, Each
Quantity
Packaging
Catalog number: 442730050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75178-96-0
IUPAC Name
tert-butyl N-(3-azaniumylpropyl)carbamate
Molecular Formula
C8H19N2O2
InChI Key
POHWAQLZBIMPRN-UHFFFAOYSA-O
SMILES
CC(C)(C)OC(=O)NCCC[NH3+]
Appearance (Color)
Colorless to white
Infrared spectrum
Conforms
Appearance (Form)
Liquid or low melting solid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text