Thermo Scientific Chemicals

(S)-3-Fluoropyrrolidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 442740050
5 g, Each
Thermo Scientific Chemicals

(S)-3-Fluoropyrrolidine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 442740050
5 g, Each
Quantity
Catalog number: 442740050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
136725-53-6
IUPAC Name
3-fluoropyrrolidine hydrochloride
Molecular Formula
C4H9ClFN
InChI Key
LENYOXXELREKGZ-UHFFFAOYNA-N
SMILES
Cl.FC1CCNC1
Appearance (Color)
White to light brown
Infrared spectrum
Conforms
Specific optical rotation
+5° to +11° (20°C, 589 nm) (c=1 in MeOH)
Appearance (Form)
Powder
Titration Argentometric
>=96.0 to =<104.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text