Thermo Scientific Chemicals

Methyl 4-aminobutyrate hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 442911000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

Methyl 4-aminobutyrate hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 442911000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 442911000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13031-60-2
IUPAC Name
4-methoxy-4-oxobutan-1-aminium chloride
Molecular Formula
C5H12ClNO2
InChI Key
WPGPRLVPWACBHW-UHFFFAOYSA-N
SMILES
[Cl-].COC(=O)CCC[NH3+]
Titration Argentometric
98.5 to 101.5 %
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text