Thermo Scientific Chemicals

L-Alanine tert-butyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 442920010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

L-Alanine tert-butyl ester hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 442920010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 442920010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13404-22-3
IUPAC Name
tert-butyl (2S)-2-aminopropanoate hydrochloride
Molecular Formula
C7H16ClNO2
InChI Key
WIQIWPPQGWGVHD-JEDNCBNOSA-N
SMILES
Cl.C[C@H](N)C(=O)OC(C)(C)C
Appearance (Form)
Crystalline powder or solid
Titration Argentometric
>=98.5 to =<101.5 %
Appearance (Color)
White to off-white
Specific optical rotation
+1.2° to +1.6° (20°C, 589 nm) (c=2 in EtOH)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text