BOC-N-Methyl-L-alanine, 99%
BOC-N-Methyl-L-alanine, 99%
BOC-N-Methyl-L-alanine, 99%
BOC-N-Methyl-L-alanine, 99%
Thermo Scientific Chemicals

BOC-N-Methyl-L-alanine, 99%

CAS: 16948-16-6 | C9H17NO4 | 203.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 442930050
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS16948-16-6
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl](methyl)amino}propanoic acid
Molecular FormulaC9H17NO4
InChI KeyVLHQXRIIQSTJCQ-LURJTMIESA-N
SMILESC[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration with NaOH98.5 to 101.5 %
Infrared spectrumConforms
Specific optical rotation-29° to -31° (20°C, 589 nm) (c=1 in EtOH)
Appearance (Form)Powder
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
342683Certificate of AnalysisJan 31, 2025442930050, 442930010
341313Certificate of AnalysisJan 22, 2024442930050, 442930010
342683Certificate of AnalysisJul 01, 2022442930050, 442930010
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection