Thermo Scientific Chemicals

1,2-Dichloroethane, 99.5%, Thermo Scientific Chemicals

Catalog number: 443011000
100 mL, Each
Thermo Scientific Chemicals

1,2-Dichloroethane, 99.5%, Thermo Scientific Chemicals

Catalog number: 443011000
100 mL, Each
Quantity
Catalog number: 443011000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
107-06-2
IUPAC Name
1,2-dichloroethane
Molecular Formula
C2H4Cl2
InChI Key
WSLDOOZREJYCGB-UHFFFAOYSA-N
SMILES
ClCCCl
Appearance (Color)
Clear colorless
GC
>=99.4 %
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Water
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text