Thermo Scientific Chemicals

2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals

Catalog number: 443150100
10 g, Each
Thermo Scientific Chemicals

2-Mercaptoethanesulfonic acid sodium salt, 98%, analytical standard, Thermo Scientific Chemicals

Catalog number: 443150100
10 g, Each
Quantity
Catalog number: 443150100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19767-45-4
IUPAC Name
sodium 2-sulfanylethane-1-sulfonate
Molecular Formula
C2H5NaO3S2
InChI Key
XOGTZOOQQBDUSI-UHFFFAOYSA-M
SMILES
[Na+].[O-]S(=O)(=O)CCS
Appearance (Form)
Crystalline powder
Loss on drying
=<2 %
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Titration Iodometric
>=97.5 to 102 % (On dried substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text