Thermo Scientific Chemicals

Methyl pyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 443170050
5 g, Each
Thermo Scientific Chemicals

Methyl pyrazine-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 443170050
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 443170050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6164-79-0
IUPAC Name
methyl pyrazine-2-carboxylate
Molecular Formula
C6H6N2O2
InChI Key
TWIIRMSFZNYMQE-UHFFFAOYSA-N
SMILES
COC(=O)C1=CN=CC=N1
Appearance (Color)
White to yellow to light orange
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
GC
>=96.0 %
Melting point
57°C to 63°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text