Applied Biosystems™

RNaseP Verification Plate, Fast 96-well

Catalog number: 4432656
Have Questions?
Applied Biosystems™

RNaseP Verification Plate, Fast 96-well

Catalog number: 4432656
Have Questions?
Catalog Number
4432656
Unit Size
1 x 96 well plate
Price (USD)
Catalog NumberUnit SizePrice (USD)
44326561 x 96 well plate
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Fast 96-Well RNaseP Verification Plate is used to verify your real-time PCR instrument with Fast 96-Well Block. Confirm the performance of your instrument by running the RNaseP assay. Please see your instrument user guide for full instructions on use.

The Fast 96-Well RNaseP Verification Plate is preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye-labeled probe) and template.

The verification plate must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, it is recommended that the performance of your instrument be verified every six months.

Specifications

Calibrated Dye
Proprietary Mix
For Use With (Equipment)
ViiA™ 7 Dx System, QuantStudio™ Dx
Reaction Speed
Fast
Product Type
qPCR System Verification Plate
For Use With (Application)
Real Time PCR (qPCR)
Shipping Condition
Dry Ice
Calibration Type
Instrument Verification Plate
PCR Method
qPCR

Contents & Storage

Store at –20°C ± 5°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text