2-Amino-5-methoxybenzoic acid, 97%, Thermo Scientific Chemicals
2-Amino-5-methoxybenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-methoxybenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 443340050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6705-03-9
IUPAC Name2-amino-5-methoxybenzoic acid
Molecular FormulaC8H9NO3
InChI KeyUMKSAURFQFUULT-UHFFFAOYSA-N
SMILESCOC1=CC=C(N)C(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or solid
HPLC>=96.0 %
Appearance (Color)Off-white to brown to purple
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only