Thermo Scientific Chemicals

Picolinamide, 98%, Thermo Scientific Chemicals

Catalog number: 443360100
10 g, Each
Thermo Scientific Chemicals

Picolinamide, 98%, Thermo Scientific Chemicals

Catalog number: 443360100
10 g, Each
Quantity
Catalog number: 443360100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1452-77-3
Infrared spectrum
Conforms
Appearance (Form)
Powder
Appearance (Color)
White to off-white
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text