Thermo Scientific Chemicals

4-Bromo-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 443400250
25 g, Each
Thermo Scientific Chemicals

4-Bromo-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Catalog number: 443400250
25 g, Each
Quantity
Catalog number: 443400250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
68837-59-2
IUPAC Name
4-bromo-2-methylbenzoic acid
Molecular Formula
C8H7BrO2
InChI Key
RVCJOGNLYVNRDN-UHFFFAOYSA-N
SMILES
CC1=CC(Br)=CC=C1C(O)=O
Infrared spectrum
Conforms
HPLC
>=96.0 %
Appearance (Color)
White to light brown
Melting point
180°C to 185°C
Appearance (Form)
Powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text