Thermo Scientific Chemicals

4-(Trifluoromethoxy)iodobenzene, 97%, Thermo Scientific Chemicals

Catalog number: 443410010
1 g, Each
Thermo Scientific Chemicals

4-(Trifluoromethoxy)iodobenzene, 97%, Thermo Scientific Chemicals

Catalog number: 443410010
1 g, Each
Quantity
Catalog number: 443410010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
103962-05-6
IUPAC Name
1-iodo-4-(trifluoromethoxy)benzene
Molecular Formula
C7H4F3IO
InChI Key
RTUDBROGOZBBIC-UHFFFAOYSA-N
SMILES
FC(F)(F)OC1=CC=C(I)C=C1
Appearance (Form)
Liquid
GC
>=96.0 %
Appearance (Color)
Colorless to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text