Thermo Scientific Chemicals

2-Methylpyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 443490010
1 g, Each
Thermo Scientific Chemicals

2-Methylpyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 443490010
1 g, Each
Quantity
Catalog number: 443490010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4021-11-8
IUPAC Name
2-methylpyridine-4-carboxylic acid
Molecular Formula
C7H7NO2
InChI Key
PMDHIMMPXRSDML-UHFFFAOYSA-N
SMILES
CC1=NC=CC(=C1)C(O)=O
Appearance (Color)
White to brown
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or solid
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text