2-Methylpyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
2-Methylpyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylpyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 443490050
Price (USD)/ Each
357.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
357.00
Add to cart
2-Methylpyridine-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog number443490050
Price (USD)/ Each
357.00
-
Add to cart
Chemical Identifiers
CAS4021-11-8
IUPAC Name2-methylpyridine-4-carboxylic acid
Molecular FormulaC7H7NO2
InChI KeyPMDHIMMPXRSDML-UHFFFAOYSA-N
SMILESCC1=NC=CC(=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder or solid
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only