1x1 image pixel for data collection
Agar, Bacteriological grade
Agar, Bacteriological grade
Thermo Scientific Chemicals

Agar, Bacteriological grade

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Catalog number 443572500
Price (USD)
85.65
Special offer
Online exclusive
Ends: 26-Sep-2025
105.00
Save 19.35 (18%)
Each
In stock
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
Ends: 26-Sep-2025
105.00
Save 19.35 (18%)
Each
Add to cart
Chemical Identifiers
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow to beige
Ash=<5 %
Solubility(1.5 % in hot water) Clear to slightly
Infrared spectrumConforms
pH6.0 to 7.5 (10 % suspension)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Microbial media

RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection