Thermo Scientific Chemicals

Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 443780050
5 g, Each
Thermo Scientific Chemicals

Cyclopropylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 443780050
5 g, Each
Quantity
Catalog number: 443780050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5239-82-7
IUPAC Name
2-cyclopropylacetic acid
Molecular Formula
C5H8O22
InChI Key
BXXOHRWKRXBZTG-UHFFFAOYSA-N
SMILES
CN1CCN(C)B1N=S=NB1N(C)CCN1C
Appearance (Color)
Colorless to light yellow
Titration with NaOH
>=96.0 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text