Thermo Scientific Chemicals

4-Acetamidobenzenesulfonyl azide, 97%, Thermo Scientific Chemicals

Catalog number: 443800050
5 g, Each
Thermo Scientific Chemicals

4-Acetamidobenzenesulfonyl azide, 97%, Thermo Scientific Chemicals

Catalog number: 443800050
5 g, Each
Quantity
Catalog number: 443800050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2158-14-7
IUPAC Name
4-acetamidobenzene-1-sulfonyl azide
Molecular Formula
C8H8N4O3S
InChI Key
NTMHWRHEGDRTPD-UHFFFAOYSA-N
SMILES
CC(=O)NC1=CC=C(C=C1)S(=O)(=O)N=[N+]=[N-]
Appearance (Color)
White to yellow to beige to grey to brown
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
108°C to 116°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text