Thermo Scientific Chemicals

4-Amino-2-chlorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: 443880100
10 g, Each
Thermo Scientific Chemicals

4-Amino-2-chlorobenzonitrile, 99%, Thermo Scientific Chemicals

Catalog number: 443880100
10 g, Each
Quantity
Catalog number: 443880100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20925-27-3
IUPAC Name
4-amino-2-chlorobenzonitrile
Molecular Formula
C7H5ClN2
InChI Key
ZFBKYGFPUCUYIF-UHFFFAOYSA-N
SMILES
NC1=CC=C(C#N)C(Cl)=C1
Water
=<0.5 %
Appearance (Color)
White to cream to brown
HPLC
>=98.5 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text